Learn More
Ethyl-Beta-D-glucuronide, Lipomed
Description
Ethyl-Beta-D-glucuronide
Specifications
Specifications
| Chemical Name or Material | Ethyl-beta-D-glucuronide |
| CAS | 17685-04-0 |
| Analyte or Component Names | Ethyl-beta-D-glucuronide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H14 O7 |
| Synonym | β-D-Glucopyranosiduronic acid, ethyl (9 CI, ACI), Ethyl β-D-glucopyranosiduronic acid (ACI), Glucopyranosiduronic acid, ethyl, β-D- (6 CI, 8 CI), BA 2829, Ethyl glucuronide, Ethyl β-D-glucosiduronic acid, Ethyl β-D-glucuronide |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | CCO[C@@H]1 O[C@@H]([C@@H](O)[C@H](O)[C@H]1 O)C(=O)O |
| IUPAC Name | (2 S,3 S,4 S,5 R,6 R)-6-ethoxy-3,4,5-trihydroxyoxane-2-carboxylic acid |
| Industry Type | Pharmaceutical |
| Show More |