Learn More
Ethyl Benzoylformate-d5, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl Benzoylformate-d5 |
| CAS | 1025892-26-5 |
| Molecular Formula | C10H5D5O3 |
| Quantity | 250 mg |
| Synonym | alpha-Oxo-benzeneacetic-d5 Acid Ethyl Ester, Phenyl-glyoxylic-d5 Acid Ethyl Ester, 2-Oxo-2-phenylacetic-d5 Acid Ethyl Ester, Ethyl 2-Oxo-2-phenylacetate-d5, Ethyl 2-Phenyl-2-oxoacetate-d5, NSC 6766-d5, Phenylglyoxylic-d5 Acid Ethyl Ester |
| InChI Formula | InChI=1S/C10H10O3/c1-2-13-10(12)9(11)8-6-4-3-5-7-8/h3-7H,2H2,1H3/i3D,4D,5D,6D,7D |
| SMILES | [2H]c1c([2H])c([2H])c(C(=O)C(=O)OCC)c([2H])c1[2H] |
| Molecular Weight (g/mol) | 183.22 |
| Formula Weight | 183.0944 |