Learn More
Alfa Aesar™ Ethyl (4-formyl-2-methoxyphenoxy)acetate
Product Code. p-7033387
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11315569 250 mg 250mg
11325569 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11315569 Supplier Alfa Aesar™ Supplier No. H50491.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 51264-70-1
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.24
MDL Number MFCD01764358
InChI Key SVYWLZTVBMJCGG-UHFFFAOYSA-N
Synonym ethyl 2-4-formyl-2-methoxyphenoxy acetate, ethyl 4-formyl-2-methoxyphenoxy acetate, 4-formyl-2-methoxy-phenoxy-acetic acid ethyl ester, acmc-1ap2o, 4-carbethoxymethoxy-3-methoxybenzaldehyde, 2-methoxy-4-formyl-phenoxyessigsaureathylester, 4-formyl-2-methoxyphenoxy acetic acid ethyl ester, acetic acid, 2-4-formyl-2-methoxyphenoxy-, ethyl ester
PubChem CID 968872
IUPAC Name ethyl 2-(4-formyl-2-methoxyphenoxy)acetate
SMILES CCOC(=O)COC1=C(OC)C=C(C=O)C=C1
Specifications
Quantity 250 mg
Formula Weight 238.24
Chemical Name or Material Ethyl (4-formyl-2-methoxyphenoxy)acetate