missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-Pyridylacetate, TRC

Product Code. 30384789
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30384789 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30384789 Supplier Toronto Research Chemicals Supplier No. TRCE92587010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3-Pyridylacetate
CAS 39931-77-6
Molecular Formula C9H11NO2
Quantity 10 g
Synonym (3-Pyridyl)acetic Acid Ethyl Ester, 2-(Pyridin-3-yl)acetic Acid Ethyl Ester, Ethyl 2-(Pyridin-3-yl)acetate, Ethyl 3-Pyridineacetate, Ethyl 3-Pyridinylacetate, Ethyl 3-Pyridylacetate, NSC 76091, Pyridin-3-ylacetic Acid Ethyl Ester
InChI Formula InChI=1S/C9H11NO2/c1-2-12-9(11)6-8-4-3-5-10-7-8/h3-5,7H,2,6H2,1H3
SMILES CCOC(=O)Cc1cccnc1
IUPAC Name ethyl 2-pyridin-3-ylacetate
Molecular Weight (g/mol) 165.19
Formula Weight 165.079
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.