missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3-ethoxypropionate, 99%, Honeywell™

Product Code. p-7120413 Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Unit Size:
1 L
4 L
This item is not returnable. View return policy

Product Code. 15645780

Brand: Honeywell Research Chemicals 5375861L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

0.99

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 763-69-9
Molecular Formula C7H14O3
Molecular Weight (g/mol) 146.186
MDL Number MFCD00051356
InChI Key BHXIWUJLHYHGSJ-UHFFFAOYSA-N
Synonym ethyl 3-ethoxypropionate, 3-ethoxypropionic acid ethyl ester, propanoic acid, 3-ethoxy-, ethyl ester, eep solvent, ethoxypropionic acid, ethyl ester, ethyl beta-ethoxypropionate, unii-ec38rsj79j, propionic acid, 3-ethoxy-, ethyl ester, ethyl-3-ethoxypropionate, ethylester kyseliny 3-ethoxypropionove
PubChem CID 12989
IUPAC Name ethyl 3-ethoxypropanoate
SMILES CCOCCC(=O)OCC

Specifications

Melting Point 49.99°C
Color Colorless
Density 0.95g/mL at 25°C
Boiling Point 166°C
Flash Point 58°C (136.4°F)
Packaging Glass Bottle
Linear Formula C2H5OCH2CH2CO2C2H5
Refractive Index n20/D 1.405
Quantity 1 L
UN Number UN3272
Vapor Density 5.03 (vs air)
Formula Weight 146.18g/mol
Percent Purity 99%
Chemical Name or Material Ethyl 3-ethoxypropionate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.