missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 3,3,3-Trifluoropropanoate, TRC

Product Code. 30329015
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30329015 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30329015 Supplier Toronto Research Chemicals Supplier No. TRCB440230250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Ethyl 3,3,3-Trifluoropropanoate
CAS 352-23-8
Molecular Formula C5 H7 F3 O2
Quantity 250 mg
InChI Formula InChI=1S/C5H7F3O2/c1-2-10-4(9)3-5(6,7)8/h2-3H2,1H3
SMILES CCOC(=O)CC(F)(F)F
IUPAC Name ethyl 3,3,3-trifluoropropanoate
Molecular Weight (g/mol) 156.103
Formula Weight 156.04
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.