missing translation for 'onlineSavingsMsg'
Learn More

Ethyl 2-bromo-4,4,4-trifluorobutyrate, 97%

Product Code. 15455317
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. Quantity unitSize
15455317 1 g 1g
15465317 250 mg 250mg
2 options
This item is not returnable. View return policy

Product Code. 15455317

Brand: Thermo Scientific Alfa Aesar undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 367-33-9 | C6H8BrF3O2 | 249.027 g/mol

Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ethyl 2-bromo-4,4,4-trifluorobutyrate is used as a pharmaceutical intermediate.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 367-33-9
Molecular Formula C6H8BrF3O2
Molecular Weight (g/mol) 249.027
MDL Number MFCD10687235
InChI Key POVYUFTVLGOXEE-UHFFFAOYSA-N
Synonym 2-bromo-4,4,4-trifluorobutyric acid ethyl ester, ethyl 2-bromo-4,4,4-trifluorobutyrate, ethyl 2-bromo-4,4,4-trifluoro-butanoate, butanoic acid,2-bromo-4,4,4-trifluoro-, ethyl ester
PubChem CID 45594306
IUPAC Name ethyl 2-bromo-4,4,4-trifluorobutanoate
SMILES CCOC(=O)C(CC(F)(F)F)Br

Specifications

Boiling Point 67°C (18 mmHg)
Quantity 1 g
UN Number UN3265
Formula Weight 249.03
Percent Purity 97%
Chemical Name or Material Ethyl 2-bromo-4,4,4-trifluorobutyrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.