missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ethyl 1H-Imidazole-1-carboxylate, TRC
Specifications
Specifications
| Chemical Name or Material | Ethyl 1H-Imidazole-1-carboxylate |
| CAS | 19213-72-0 |
| Molecular Formula | C6H8N2O2 |
| Quantity | 1 g |
| Synonym | Etomidate Impurity 10 (Ethyl 1H-imidazole-1-carboxylate) |
| InChI Formula | InChI=1S/C6H8N2O2/c1-2-10-6(9)8-4-3-7-5-8/h3-5H,2H2,1H3 |
| SMILES | CCOC(=O)n1ccnc1 |
| IUPAC Name | ethyl imidazole-1-carboxylate |
| Molecular Weight (g/mol) | 140.14 |
| Formula Weight | 140.0586 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?