Learn More
Ethanol Absolute Ph.Eur., Solveco
Product Code. 12697213
Click to view available options
Quantity:
1 L
5 L
25 L
Unit Size:
25 L
5 L
Pack of 12
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Product Code. 12697213
Brand: Solveco Chemicals 1016
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Regulatory: This product is subject to excise duty on alcohol in accordance with European Directive 92/83/EEC, all customers must have an authorization before delivery.
Absolute, pharmaceutical-grade ethanol with a concentration of 99.9 vol - %. This synthetic ethanol is produced through a complex three-step process, resulting in a double-distilled ethanol.
A synthetic ethanol that is produced through a complex three-step process, resulting in double-distilled ethanol. This makes it an ideal product for applications requiring the highest quality ethanol. Synthetic ethanol is manufactured using selective synthetic chemistry, which eliminates risks associated with contaminants such as genetically modified organisms, pesticide residues, natural allergens, phthalates, and dioxins. With a purity of 99.9%, this ethanol meets the quality requirements of the European Pharmacopoeia and is available with a batch-specific Certificate of Analysis upon request.
Chemical Identifiers
CAS 64-17-5
Molecular Formula C2H6O
Molecular Weight (g/mol) 46.07
InChI Key LFQSCWFLJHTTHZ-UHFFFAOYSA-N
IUPAC Name ethanol
SMILES CCO
Specifications
Boiling Point 78°C
Quantity 5 L
Packaging HDPE Container
Percent Purity 99.90%
Chemical Name or Material Ethyl Alcohol
compliance-icons
Product Identifier
  • Ethanol 99,5%
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P243-Take precautionary measures against static discharge.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.