Learn More
Cytiva ETFE Ferrule
Product Code. 11300262 Shop All Cytiva Products
Click to view available options
Diameter (English) Outside:
1/16 in.
1/8 in.
3/16 in.
Unit Size:
Each
Product Code. 11300262
Brand: Cytiva 18112706
Ferrules used for chromatography tubing. Cytiva Lifescience™ ETFE Ferrule is made from ethylenetetra fluoroethylene and comes with different outside diameters.
  • Ferrules
  • Chromatography accessory
  • Used for: protecting the end of tubing
  • Packsize: 10 Connections
  • Ethylenetetra fluoroethylene (ETFE) 
TRUSTED_SUSTAINABILITY
Specifications
Type Ferrule
Diameter (English) Outside 1/16 in.
For Use With (Equipment) Tubing
Material ETFE
Quantity 10 Pack