Learn More
Invitrogen™ Estrogen Receptor beta Polyclonal Antibody

Description
PA1-310B has been successfully used in Western blot, immunohistochemistry, immunofluorescence, and immunocytochemistry procedures. By Western blot, this antibody detects an ~55 kDa protein representing ER beta from both rat brain homogenate as well as various breast cancer cell lines. Immunocytochemical staining of ER beta in cells overexpressing the human protein results in nuclear staining. The PA1-310B immunogen is a synthetic peptide corresponding to residues C(467) S S T E D S K N K E S S Q N L Q S Q(485) of rat ER beta. This immunizing peptide (PEP-007) is available for use in neutralization and control experiments. Recombinant ER beta expressed in baculovirus system (Cat. # RP-311 and RP-312) can be purchased for control experiments in Western blot and gel shift assays.
Specifications
Specifications
| Antigen | Estrogen Receptor beta |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1mg/mL BSA and 0.05% sodium azide |
| Gene | ESR2 |
| Gene Accession No. | O08537, Q62986, Q92731 |
| Gene Alias | ER; ER beta; ER[b]; Erb; Erb2; Erbeta; ER-beta; Esr2; ESRB; ESR-BETA; ESTRB; estrogen receptor 2; estrogen receptor 2 (beta); estrogen receptor 2 (ER beta); estrogen receptor 2 beta; estrogen receptor beta; estrogen receptor beta 1 isoform deletion variant; estrogen receptor beta 4; NR3A2; Nuclear receptor subfamily 3 group A member 2; oestrogen receptor beta; steroid receptor |
| Gene Symbols | ESR2 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.