missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ERO1LB Polyclonal Antibody
GREENER_CHOICE

Product Code. 16626554
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16626554 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16626554 Supplier Invitrogen™ Supplier No. PA5100065

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ERO1LB.

ERO1LB is essential oxidoreductase that oxidizes proteins in the endoplasmic reticulum to produce disulfide bonds. ERO1LB acts by oxidizing directly P4HB/PDI isomerase through a direct disulfide exchange. ERO1LB does not act as a direct oxidant of folding substrate, but relies on P4HB/PDI to transfer oxidizing equivalent.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ERO1LB
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Ero1b
Gene Accession No. Q86YB8, Q8R2E9
Gene Alias 1300013B24Rik; 1700065B09Rik; AI447560; endoplasmic oxidoreductase 1 beta; endoplasmic oxidoreductin-1-like protein B; endoplasmic reticulum oxidoreductase 1 beta; endoplasmic reticulum oxidoreductase beta; Endoplasmic reticulum oxidoreductin-1-like protein B; Ero1b; Ero1beta; ero1-beta; ERO1LB; ERO1-L-beta; ERO1-like beta; ERO1-like beta (S. cerevisiae); ERO1-like protein beta; Oxidoreductin-1-L-beta; RGD1563548
Gene Symbols Ero1b, Ero1lb
Host Species Rabbit
Immunogen A synthesized peptide derived from human ERO1B(Accession Q86YB8), corresponding to amino acid residues W196-F246.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 364755, 56605, 67475
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.