Learn More
Lenz Laborglasintrumente Erlenmeyer Flasks Wide Neck
Product Code. p-7222497
Click to view available options
Capacity (Metric):
25 mL
100 mL
200 mL
1 L
2 L
Unit Size:
Pack of 10
Product Code. 10060451
Brand: Lenz Laborglasintrumente 07025073
Only null left
Add to basket
Add to basket
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
  • Acc. to DIN ISO 24450 and in addition to standard
  • With beaded rim
  • With graduation
  • Material: DURAN™
Specifications
Diameter (Metric) Outer Neck 72 mm
Capacity (Metric) 2 L
Includes Beaded rim and graduation
No. per Pack 10/Pk.
Diameter (Metric) Outer 153 mm