Learn More
Eriochrome Cyanine R, Pure, C.I.43820, Metal Indicator, Fisher Chemical
Shop All Fisher Chemical Products
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| CAS | 3564-18-9 |
| Molecular Formula | C23H15Na3O9S |
| MDL Number | 12411 |
| Formula Weight | 536.39g/mol |
| Packaging | Glass Jar |
| Color | Brown/Red |
| Quantity | 25 g |
| Physical Form | Solid |