Learn More
Eppendorf epT.I.P.S.™ Motion as SafeRack tips
Shop All Eppendorf Products
Click to view available options
Volume (Metric):
1.000 μL
50 μL
300 μL
Sterility:
Non-sterile
PCR Clean
Filtered:
Filtered
Non-filtered
Unit Size:
Pack of 960
Description
- Specially designed for use on the epMotion™ 5070 and 5075 Liquid Handling Workstation. epT.I.P.S. Motion SafeRack pipette tips, with and without filters
- Developed in torsion-resistant, contamination-free racks that are recommended for the re-use of tips feature of epMotion 5073 systems.
Reuse Tips Feature:
- The new epMotion systems can reuse tips as e.g. economical choice for multiple wash steps of the same well
- Pipette tips will be placed back into the position of the box they were removed from
- Returned tips will be reused within an application for the same source vessel
- Individual compartments in rack to avoid cross-contamination between used tips
- Available as standard and PCR clean quality for 50, 300, and 1000 μL
Specifications
Specifications
| Volume (Metric) | 50 μL |
| Filtered | Filtered |
| Sterility | PCR Clean |
| Color | Clear |
| Autoclavable | Not Autoclavable |
| Format | Box |
| Tip Style | Standard, Straight |
| Product Type | Robotic Tips |
| Disposable | Single-use |
| For Use With (Equipment) | Eppendorf epMotion |
| Show More |