Learn More
epsilon-Viniferin, TRC
Specifications
Specifications
| Chemical Name or Material | Epsilon-Viniferin |
| CAS | 62218-08-0 |
| Molecular Formula | C28 H22 O6 |
| Quantity | 10 mg |
| Synonym | 1,3-Benzenediol, 5-[(2R,3R)-2,3-dihydro-6-hydroxy-2-(4-hydroxyphenyl)-4-[(1E)-2-(4-hydroxyphenyl)ethenyl]-3-benzofuranyl]-, 1,3-Benzenediol, 5-[2,3-dihydro-6-hydroxy-2-(4-hydroxyphenyl)-4-[2-(4-hydroxyphenyl)ethenyl]-3-benzofuranyl]-, [2R-[2α,3β,4(E)]]- |
| InChI Formula | InChI=1S/C28H22O6/c29-20-7-2-16(3-8-20)1-4-18-11-24(33)15-25-26(18)27(19-12-22(31)14-23(32)13-19)28(34-25)17-5-9-21(30)10-6-17/h1-15,27-33H/b4-1+/t27-,28+/m1/s1 |
| SMILES | Oc1ccc(\C=C\c2cc(O)cc3O[C@H]([C@H](c4cc(O)cc(O)c4)c23)c5ccc(O)cc5)cc1 |
| IUPAC Name | 5-[(2R,3R)-6-hydroxy-2-(4-hydroxyphenyl)-4-[(E)-2-(4-hydroxyphenyl)ethenyl]-2,3-dihydro-1-benzofuran-3-yl]benzene-1,3-diol |
| Molecular Weight (g/mol) | 454.471 |
| Formula Weight | 454.142 |