missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EPS8L2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16650955 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16650955 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16650955 Supplier Invitrogen™ Supplier No. PA5104504

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ES8L2.

The protein encoded by this gene has been shown to be involved in the regulation of cell proliferation, and in the crosstalk between the adiponectin signalling and insulin signalling pathways. The encoded protein binds many other proteins, including RAB5A, DCC, AKT2, PIK3CA, adiponectin receptors, and proteins of the NuRD/MeCP1 complex. This protein is found associated with endosomal membranes, but can be released by EGF and translocated to the nucleus.
TRUSTED_SUSTAINABILITY

Specifications

Antigen EPS8L2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EPS8L2
Gene Accession No. Q99K30, Q9H6S3
Gene Alias AI042819; AW545405; Epidermal growth factor receptor kinase substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-like protein 2; epidermal growth factor receptor pathway substrate 8-related protein 2; EPS8 like 2; EPS8 related protein 2; Eps8l2; EPS8-like 2; EPS8-like protein 2; EPS8R2; EPS8-related protein 2; PP13181
Gene Symbols EPS8L2
Host Species Rabbit
Immunogen A synthesized peptide derived from human EPS8L2(Accession Q9H6S3), corresponding to amino acid residues C10-F60.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 361674, 64787, 98845
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.