missing translation for 'onlineSavingsMsg'
Learn More

Eprenetapopt, MedChemExpress

Product Code. p-200100227
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16480458

Brand: Medchem Express HY19980

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. Eprenetapopt triggers apoptosis in tumor cells. Eprenetapopt also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance.

Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. Eprenetapopt triggers apoptosis in tumor cells. Eprenetapopt also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance.

Specifications

Chemical Name or Material Eprenetapopt
CAS 5291-32-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H17NO3
Quantity 5 mg
Synonym APR-246 PRIMA-1Met
Solubility Information DMSO : 100 mg/mL (501.88 mM; Need ultrasonic) ∣H2O : 50 mg/mL (250.94 mM; Need ultrasonic)
SMILES O=C1C(COC)(CO)N2CCC1CC2
Molecular Weight (g/mol) 199.25
Formula Weight 199.25
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.