Learn More
Eprenetapopt, MedChemExpress
Product Code. 16480458
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16480458
Brand: Medchem Express HY19980
MedChemExpress Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. Eprenetapopt triggers apoptosis in tumor cells. Eprenetapopt also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance.
Eprenetapopt (APR-246) is a first-in-class, small molecule that restores wild-type p53 functions in TP53-mutant cells. Eprenetapopt triggers apoptosis in tumor cells. Eprenetapopt also targets the selenoprotein thioredoxin reductase 1 (TrxR1), a key regulator of cellular redox balance.
Specifications
Chemical Name or Material Eprenetapopt
CAS 5291-32-7
Color Off-White
Purity Grade Notes Research
Molecular Formula C10H17NO3
Quantity 5 mg
Synonym APR-246 PRIMA-1Met
Solubility Information DMSO : 100 mg/mL (501.88 mM; Need ultrasonic) ∣H2O : 50 mg/mL (250.94 mM; Need ultrasonic)
SMILES O=C1C(COC)(CO)N2CCC1CC2
Molecular Weight (g/mol) 199.25
Formula Weight 199.25
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death