Learn More
Eppendorf™ Space Saving Dolly for Innova 3100 Shakers
Description
- Color: Black
- Four wheels
- Dimensions: 71.76L × 45.72cmW (28.25 × 18 in.)
Specifications
Specifications
| Product Type | Space-saving Dolly |
| Height (English) | 20 in. |
| Height (Metric) | 51 cm |
| Length (English) | 28.25 in. |
| Length (Metric) | 71.75 cm |
| Width (Metric) | 45.72 cm |
| For Use With (Equipment) | Innova 3100 High-temp Water Bath Shaker |