Learn More
Eppendorf™ S-4-104 Rotor Adapters
Product Code. p-4501295 Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 250 mL Tubes
1 x 500 mL Bottle
14 x 15 mL Conical Tubes
14 x 5 mL Conical Tubes
20 x 5.5/12 mL Tubes
23 x 2.6/8 mL Tubes
23 x 7/17 mL Tubes
4 x 750 mL Wide-mouth Bottles
5 x 50 mL Conical Tubes
50 x 1.5/2 mL Tubes
7 x 50 mL Conical Tubes
Unit Size:
1 set
Pack of 2
Product Code. 13256619
Brand: Eppendorf™ 5825732006
Includes
2 piece set
Available in a wide range of configurations to suit a variety of applications.
  • For use with Centrifuge 5810/5810 R
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Capacity 5 x 50 mL Conical Tubes
For Use With (Equipment) S-4-104 rotor for 5810/5810R centrifuge
No. per Pack 2 Pack