Learn More
Eppendorf™ A-4-38 Rotor Adapters
Shop All Eppendorf Products
Click to view available options
Capacity:
1 x 5 mL Conical Tube
9 x 5 mL Tubes
Unit Size:
Pack of 2
Description
- For use with Centrifuge 5702/ 5702 R/5702 RH and A-4-38 Rotor
- Designed to meet the requirement of low- to mid-throughput clinical research and cell culture labs.
Specifications
Specifications
| Type | Rotor Adapter |
| Capacity | 9 x 5 mL Tubes |
| For Use With (Equipment) | A-4-38 |