Learn More
epi-Canagliflozin, TRC
Specifications
Specifications
| Chemical Name or Material | epi-Canagliflozin |
| CAS | 1589590-87-3 |
| Molecular Formula | C24H25FO5S |
| Quantity | 25 mg |
| Synonym | (1R)-1,5-Anhydro-1-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]-D-glucitol |
| InChI Formula | InChI=1S/C24H25FO5S/c1-13-2-3-15(24-23(29)22(28)21(27)19(12-26)30-24)10-16(13)11-18-8-9-20(31-18)14-4-6-17(25)7-5-14/h2-10,19,21-24,26-29H,11-12H2,1H3 |
| SMILES | OCC1O[C@@H](C2=CC(CC3=CC=C(C4=CC=C(F)C=C4)S3)=C(C)C=C2)C(O)C(O)[C@@H]1O |
| IUPAC Name | 2-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]-6-(hydroxymethyl)oxane-3,4,5-triol |
| Molecular Weight (g/mol) | 444.52 |
| Formula Weight | 444.1407 |