missing translation for 'onlineSavingsMsg'
Learn More

Eosin B

Product Code. p-7023627
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11413627

Brand: Thermo Scientific Alfa Aesar A17377.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 548-24-3 | C20H6Br2N2Na2O9

Eosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Eosin B sodium salt is a cell and tissue stain. Eosin B sodium salt is used as a counter stain for collagen, as a tissue stain for cell granules, nuclei, and microorganisms when coupled with Azure A (sc-203729), and as a stain for cell differentiation of the anterior pituitary.

Solubility
Soluble in water (390 g/L at 20°C), and ethanol.

Notes
Store at room temperature. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-24-3
Molecular Formula C20H6Br2N2Na2O9
MDL Number MFCD00005041
Synonym C.I. 45400; 4,5-Dibromo-2,7-dinitrofluorescein disodium salt

Specifications

Quantity 25 g
Solubility Information Soluble in water (390g/L at 20°C),and ethanol.
Formula Weight 624.08
Chemical Name or Material Eosin B

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.