missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eosin B, Free Acid, MP Biomedicals
Shop All MP Biomedicals Products
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| CAS | 56360-46-4 |
| Assay Percent Range | ∽ 85% |
| Molecular Formula | C20H6Br2N2O9-2 |
| Synonym | C.I. 45400 |
| InChI Key | ZBQZBWKNGDEDOA-UHFFFAOYSA-L |
| SMILES | C1=CC=C2C(=C1)C(=O)OC23C4=CC(=C(C(=C4OC5=C(C(=C(C=C35)[N+](=O)[O-])[O-])Br)Br)[O-])[N+](=O)[O-] |
| IUPAC Name | 4',5'-dibromo-2',7'-dinitro-3-oxospiro[2-benzofuran-1,9'-xanthene]-3',6'-diolate |
| Molecular Weight (g/mol) | 578.081 |
| PubChem CID | 7058116 |
| ChEBI | CHEBI:68617 |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction