missing translation for 'onlineSavingsMsg'
Learn More
Learn More
EOMES Antibody (1219A), mFluor Violet 610 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB8889MFV610
This item is not returnable.
View return policy
Description
EOMES Monoclonal antibody specifically detects EOMES in Mouse samples. It is validated for Flow Cytometry
Specifications
| EOMES | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Innate Immunity | |
| 8320 | |
| Store at 4C in the dark. |
| Flow Cytometry | |
| 1219A | |
| Flow Cytometry | |
| eomesodermin, eomesodermin (Xenopus laevis) homolog, t box, brain, 2, T-box brain protein 2, T-box brain2, TBR-2, TBR2eomesodermin homolog, T-brain-2 | |
| E. coli-derived recombinant mouse EOMES, Met1-Ser126, Accession # O54839 | |
| RUO | |
| Primary | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction