Learn More
Memmert™ Entry Port 23 mm for UF1060plus, Standard Position Left Side Center/Top
Product Code. 17598855 Shop All Memmert Products
Click to view available options
Diameter (Metric):
23 mm
Unit Size:
Each
Product Code. 17598855
Brand: Memmert™ UF1060plusF1
Only null left
Add to basket
Add to basket
Optional entry port for Memmert Ovens.
  • Entry port with 23 mm diameter
  • Standard position left side center/top
Specifications
For Use With (Equipment) UF1060plus Oven
Type Entry Port
Diameter (Metric) 23 mm