Learn More
Memmert™ Entry Port 23 mm, Flap & Stopper Closure, Standard Position Right Side Centre/Centre, For Un55Pa
Product Code. 16354447 Shop All Memmert Products
Click to view available options
Diameter (English):
0.9 in.
Unit Size:
Each
Product Code. 16354447
Brand: Memmert™ UN55paF2
Only null left
Add to basket
Add to basket
Use these entry ports with convection ovens. Memmert™ Convection and Universal Oven Entry Points can be closed by flap in special positions at the back.
  • Entry port with clear diameter
  • For Memmert Ovens
Specifications
For Use With (Equipment) Models 30, 55, 75, 110, 160, 260, 450, 750, 1060
Type Entry Port
Material Stainless Steel
Diameter (English) 0.9 in.
Diameter (Metric) 23 mm