missing translation for 'onlineSavingsMsg'
Learn More

ent-Efavirenz, TRC

Product Code. 30369367
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369367 2.5 mg 2.5mg
30369138 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30369367 Supplier Toronto Research Chemicals Supplier No. TRCE4250052.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154801-74-8
Molecular Formula C14 H9 Cl F3 N O2
Molecular Weight (g/mol) 315.68
IUPAC Name (4R)-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one
SMILES FC(F)(F)[C@@]1(OC(=O)Nc2ccc(Cl)cc12)C#CC3CC3

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C14H9ClF3NO2/c15-9-3-4-11-10(7-9)13(14(16,17)18,21-12(20)19-11)6-5-8-1-2-8/h3-4,7-8H,1-2H2,(H,19,20)/t13-/m1/s1
Formula Weight 315.0274
Chemical Name or Material ent Efavirenz
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.