missing translation for 'onlineSavingsMsg'
Learn More

Scharlab Enrichment Enterobacteriaceaes Broth (EE Broth)

Product Code. 11411787
Click to view available options
:
Enrichment Enterobacteriaceaes Broth; Form: Liquid
This item is not returnable. View return policy

Product Code. 11411787

Brand: Scharlab 02064500

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Enrich. Enterobacteriaceae B. (EE Broth) is used for the enrichment of enterobacteria.

  • EP/HP/ISO/USP
  • Liquid culture medium used for the enrichment of enterobacteria
  • According to the ISO 21528-1 standard and the Pharmacopeial Harmonised Method

Specifications

Certifications/Compliance ISO 21528-1
Form Liquid
Media Type Enrichment Enterobacteriaceaes Broth
Product Type Microbiology Media
Target Organism Class Gram-negative Bacteria
Formulation Enzymatic Digest of Animal Tissues (Gelatin Peptone) 10.000 g/L, Dextrose 5.000 g/L, Ox Bile 20.000 g/L, Disodium Phosphate (Anhy.) 6.450 g/L, Potassium Dihydrogenphosphate 2.000 g/L, Brilliant Green 0.015 g/L
For Use With (Application) Enrichment of Enterobacteria
Incubation Conditions 30°C to 35°C
Incubation Time 24 to 48 hr.
pH 7.2 ±0.2
Quantity 500 g
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.