missing translation for 'onlineSavingsMsg'
Learn More

(±)-Enitociclib, MedChemExpress

Product Code. 16476497
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476497

Brand: Medchem Express HY103019A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (±)-Enitociclib ((±)-BAY-1251152) is a racemic mixture of BAY-1251152. BAY-1251152 is a potent and highly selective PTEF/CDK9 inhibitor.

(±)-Enitociclib ((±)-BAY-1251152) is a racemic mixture of BAY-1251152. BAY-1251152 is a potent and highly selective PTEF/CDK9 inhibitor.

Specifications

Chemical Name or Material (±)-Enitociclib
CAS 1610358-53-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H18F2N4O2S
Quantity 5 mg
Synonym (±)-BAY-1251152 (±)-VIP152
Solubility Information DMSO : 50 mg/mL (123.63 mM; Need ultrasonic)
SMILES N=S(CC1=CC(NC2=NC=C(F)C(C3=CC=C(F)C=C3OC)=C2)=NC=C1)(C)=O
Molecular Weight (g/mol) 404.43
Formula Weight 404.43
Percent Purity 99.87%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.