missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eniporide hydrochloride, MedChemExpress
Description
Eniporide hydrochloride (EMD-96785 hydrochloride) is a potent Na+/H+ exchange inhibitor.
Specifications
Specifications
| Chemical Name or Material | Eniporide hydrochloride |
| CAS | 211813-86-4 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C14H17ClN4O3S |
| Quantity | 5 mg |
| Synonym | EMD-96785 hydrochloride |
| Solubility Information | DMSO : 55 mg/mL (154.14 mM; Need ultrasonic) ∣H2O : 4.17 mg/mL (11.69 mM; ultrasonic and warming and heat to 60°C) |
| SMILES | CS(=O)(C1=C(N2C=CC=C2)C=C(C)C(C(NC(N)=N)=O)=C1)=O.[H]Cl |
| Molecular Weight (g/mol) | 356.83 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?