missing translation for 'onlineSavingsMsg'
Learn More

Emtricitabine Triphosphate Triethylammonium Salt, TRC

Product Code. 30009649
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009649 1 mg 1mg
30008566 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009649 Supplier Toronto Research Chemicals Supplier No. TRCE5250751MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8H13FN3O12P3S •3(C6H15N)
Molecular Weight (g/mol) 487.19101193
Synonym Triphosphoric Acid P-[[(2R,5S)-5-(4-Amino-5-fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3- oxathiolan-2-yl]methyl] Ester Triethylammonium Salt, Triphosphoric Acid (2R-cis)-P-[[5-(4-Amino-5- fluoro-2-oxo-1(2H)-pyrimidinyl)-1,3-oxathiolan-2-yl]methyl] Ester Triethylammonium Salt, Emtricitabine 5'-Triphosphate Triethylammonium Salt, (-)-FTC-Triphosphate
SMILES [C]CN(CC)CC.NC1=NC(N([C@@H]2CSC(COP(OP(O)(OP(O)(O)=O)=O)(O)=O)O2)C=C1F)=O

Specifications

Analyte or Component Names Emtricitabine Triphosphate Triethylammonium Salt
Shipping Condition Blue Ice
InChI Formula InChI=1S/C8H13FN3O12P3S.C6H15N/c9-4-1-12(8(13)11-7(4)10)5-3-28-6(22-5)2-21-26(17,18)24-27(19,20)23-25(14,15)16;1-4-7(5-2)6-3/h1,5-6H,2-3H2,(H,17,18)(H,19,20)(H2,10,11,13)(H2,14,15,16);4-6H2,1-3H3/t5-,6?;/m0./s1
Solution Type Neat
Industry Type Forensics
Quantity 1 mg
Chemical Name or Material Emtricitabine Triphosphate Triethylammonium Salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.