Learn More
BD Emerald™ Sterile 2mL Syringe with 22G Needle
Description
Advantages of BD Emerald™:
- Eco-conscious manufacturing process uses solar and wind energy
- Reduced environmental footprint by using fewer raw materials
- More efficient use of raw materials limits waste and disposal costs
- Better visualization, thanks to the green piston seal
Specifications
Specifications
| Material | Polypropylene |
| Color | Clear |
| Needle Gauge | 22 G |
| Includes | Central cone |
| Length (English) Needle | 1.25 in. |
| Disposable | Single-use |
| Graduations | 0.01 mL |
| Model | Emerald |
| Quantity | 100 Pk. |
| Packaging | Individually Wrapped |
| Show More |