Learn More
Merck MultiScreenHTS 96-Well Filter Plates
Product Code. 10492352 Shop All Merck Products
Change view
Click to view available options
Pore Size:
0.45 μm
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pore Size unitSize
10492352 0.45 μm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10492352 Supplier Merck Supplier No. MSIPS4510
Only null left
Add to basket
Add to basket
For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation.
  • High Throughput
  • Meet ANSI/SBS 2004 plate specifications
  • Protects underdrain, eliminates surface contact with individual well ‘drip directors’
  • Plastic skirt enables stacking directly with collection plates
Applications
  • Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
  • Radioisotope detection
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance ANSI, SBS
Format Standard
Membrane Hydrophobic PVDF
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Material ABS Plastic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28cm2
For Use With (Application) ELISPOT assays
Quantity 10/Pk.