Learn More
Merck MultiScreen™ Centrifuge Alignment Frame
Shop All Merck Products
Click to view available options
Quantity:
4/Pk.
Unit Size:
Pack of 4
Description
- Transfer samples filtrate to the wells of a 96-well collection plate
- Prevents spillage or cross-contamination
Specifications
Specifications
| Color | Blue |
| Material | ABS Plastic |
| Sterility | Non-sterile |
| For Use With (Application) | Use to align MultiScreen plate to 96-well receiver plate for centrifugation of aqueous media |
| Quantity | 4/Pk. |