Learn More
Merck MultiScreen™ 96-Well Assay Plates for ELISPOT Assays
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
Brand:  Merck MSIPN4W50
1519.94 EUR valid until 2024-12-31
Use promo code "23269" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10307473
  • 1655.00 EUR / Pack of 50
Estimated Shipment: 31-05-2024
Description
Give optimal results in immunological ELISPOT (enzyme-linked immunospot) assays by providing high protein-binding capacity with low background staining
  • Membrane-to-plate seal ensures no reagent crosstalk and limited optical crosstalk Patented underdrain support structure allows liquid volumes to be maintained for incubation times of up to 21 days
Specifications
Specifications
Standard
MultiScreen-IP Filter Plate with Immobilon™-P membrane
0.45 μm
ABS Plastic
Non-sterile
ELISPOT assays
Hydrophobic PVDF
8 x 12
White
96
0.28cm2
50/Pk.