Learn More
Merck Millipore EZ-Fit™ Manifold 6-Place for Rubber Stopper Tulip
For use in general microbiological analysis
Brand:  Merck Millipore EZFITH0LD6
Product Code. 14872021
  • ONLINE EXCLUSIVE PRICE
    7100.00 EUR
    5030.00 EUR / Each
    Save 2070.00 EUR
    29% Off
Only null left
Estimated Shipment: 23-01-2026
Description
  • Vacuum manifolds
  • Support simultaneous filtration of six test samples
  • Made from stainless steel
  • Available with Microfil™ head for use with presterilized, ready-to-use funnels and membranes