Learn More
Merck Accessories for XX2702550 1225 Sampling Manifold
Product Code. p-109521 Shop All Merck Products
Click to view available options
Type:
Ball Valve
Chamber Replacement Kit
O-Ring
Sample Cup Extension Barrel
Support Plate
Unit Size:
Each
Pack of 12
Pack of 30
Product Code. 10186470
Brand: Merck XX2702509
Only null left
Add to basket
Add to basket
Accessories for filter holders for sample preparation.
Accessories for filter holders for sample preparation.
TRUSTED_SUSTAINABILITY
Specifications
Autoclavable Not Autoclavable
For Use With (Equipment) EMD Millipore 1225 sampling vacuum manifold
Material Silicone
Sterility Non-sterile
Type O-Ring
Diameter (English) 0.25 in.
Diameter (Metric) Inside 6.3 mm