Learn More
Merck Accessories for XX2702550 1225 Sampling Manifold
Accessories for filter holders for sample preparation.
Brand: Merck XX2702509
Additional Details : Weight : 0.10000kg
Description
Accessories for filter holders for sample preparation.Specifications
Not Autoclavable | |
EMD Millipore 1225 sampling vacuum manifold | |
Non-sterile | |
0.25 in. |
O-ring | |
Silicone | |
Laboratory Filter Holders or Cyclones | |
6.3 mm |