missing translation for 'onlineSavingsMsg'
Learn More

Emapunil, MedChemExpress

Product Code. 16459167
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize'
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16459167

missing translation for 'mfr': Medchem Express HY15527

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Emapunil (AC-5216), an orally active and selective TSPO (a mitochondrial benzodiazepine receptor) ligand, produces anti-anxiety and antidepressant-like effects in various animal models.

Emapunil (AC-5216), an orally active and selective TSPO (a mitochondrial benzodiazepine receptor) ligand, produces anti-anxiety and antidepressant-like effects in various animal models.

Specifications

Chemical Name or Material Emapunil
CAS 226954-04-7
Color White
Purity Grade Notes Research
Molecular Formula C23H23N5O2
Quantity 5 mg
Synonym AC-5216 XBD-173
Solubility Information DMSO : 33.33 mg/mL (83.02 mM; Need ultrasonic)
SMILES O=C(N(CC)CC1=CC=CC=C1)CN2C(N(C)C3=CN=C(C4=CC=CC=C4)N=C23)=O
Molecular Weight (g/mol) 401.46
Formula Weight 401.46
Percent Purity 99.26%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.