missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ELQ-300, MedChemExpress
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Description
ELQ-300 is a potent and orally bioavailable antimalarial agent, acts as an inhibitor of the reductive (Qi) site of the cytochrome bc1 complex (cyt bc1).
Specifications
Specifications
| Chemical Name or Material | ELQ-300 |
| Physical Form | Solid |
| CAS | 1354745-52-0 |
| Color | Off-White |
| Quantity | 5 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C24H17ClF3NO4 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | DMSO : 15.62 mg/mL (32.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble) |
| SMILES | O=C1C(C2=CC=C(OC3=CC=C(OC(F)(F)F)C=C3)C=C2)=C(C)NC4=C1C=C(Cl)C(OC)=C4 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction