Learn More
ELQ-300, MedChemExpress
Product Code. 16458527
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. 16458527
Brand: Medchem Express HY138365mg
MedChemExpress ELQ-300 is a potent and orally bioavailable antimalarial agent, acts as an inhibitor of the reductive (Qi) site of the cytochrome bc1 complex (cyt bc1).
ELQ-300 is a potent and orally bioavailable antimalarial agent, acts as an inhibitor of the reductive (Qi) site of the cytochrome bc1 complex (cyt bc1).
Specifications
Chemical Name or Material ELQ-300
Physical Form Solid
CAS 1354745-52-0
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C24H17ClF3NO4
For Use With (Application) COVID-19-immunoregulation
Solubility Information DMSO : 15.62 mg/mL (32.83 mM; Need ultrasonic) ∣H2O : < 0.1 mg/mL (insoluble)
SMILES O=C1C(C2=CC=C(OC3=CC=C(OC(F)(F)F)C=C3)C=C2)=C(C)NC4=C1C=C(Cl)C(OC)=C4
Molecular Weight (g/mol) 475.84
Formula Weight 475.84
Percent Purity 98.0%
Grade Research