missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ELP5 Polyclonal Antibody

Product Code. 17864590
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17864590 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17864590 Supplier Invitrogen™ Supplier No. PA5140075

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FFRDPLNWSK TEEAFPGGPL GALRAMCKRT DPVPVTIALD SLSWLLLRLP CTTLCQVLHA VSHQDSCPGD SSSVGKVSVL G Highest antigen sequence indentity to the following orthologs: Mouse - 69%, Rat - 61%.

C17orf81 belongs to the ELP5 family. C17orf81 may be involved in TP53-mediated transcriptional regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ELP5
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ELP5
Gene Accession No. Q8TE02
Gene Alias C17orf81; dermal papilla derived protein 6; Dermal papilla-derived protein 6; Dermal papilla-derived protein 6 homolog; Derp6; elongator acetyltransferase complex subunit 5; elongator complex protein 5; Elp5; HSPC002; MST071; MSTP071; Rai12; retinoic acid induced 12; retinoic acid-induced protein 12; S-phase 2 protein
Gene Symbols ELP5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ELP5. Recombinant protein control fragment (Product #RP-108893).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23587
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.