missing translation for 'onlineSavingsMsg'
Learn More

ELN-441958, MedChemExpress

Product Code. 16476298
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16476298

Brand: Medchem Express HY15043

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ELN-441958 is a potent, neutral antagonist of B1 receptor, inhibits the binding of the B1 agonist ligand [3H]DAKD to IMR-90 cells with Ki of 0.26 nM.

ELN-441958 is a potent, neutral antagonist of B1 receptor, inhibits the binding of the B1 agonist ligand [3H]DAKD to IMR-90 cells with Ki of 0.26 nM.

Specifications

Chemical Name or Material ELN-441958
CAS 913064-47-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H29ClN4O2
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (199.59 mM; Need ultrasonic)
SMILES ClC1=C2C(CN(C3=CC=CC(C(N4CCC5(CCN(C6=CC=NC=C6)CC5)CC4)=O)=C3)C2=O)=CC=C1
Molecular Weight (g/mol) 501.02
Formula Weight 501.02
Percent Purity 99.84%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.