Learn More
Fisherbrand™ PTFE-Coated Rare Earth Core Stir Bars, Elliptical
Shop All Fisherbrand Products
missing translation for 'orderingAttributeHoverText'
Length (Metric):
10 mm
15 mm
25 mm
70 mm
Diameter (Metric):
6 mm
10 mm
14 mm
28 mm
missing translation for 'unitSize'
Each
Pack of 10
Pack of 5
Description
- Use rare earth magnets for significantly increased magnetic strength compared to those using AlNiCo magnets
- Samarium-cobalt core is resistant to demagnetization
- Identified by a chemically inert, carbon black spot
Specifications
Specifications
| Autoclavable | Autoclavable |
| Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
| Color | White |
| Description | PTFE encapsulated Samarium-Cobalt magnet core |
| For Use With (Equipment) | Containers & Vessels / Stirrer Hot Plates |
| Diameter (Metric) | 14 mm |
| Length (Metric) | 25 mm |
| Quantity | 2/Pk. |
| Shape | Elliptical |