Learn More
Bel-Art™ SP Scienceware™ Ellipso-Spoon™ Spatula
Shop All SP Scienceware Products
Click to view available options
Capacity (Metric):
10 mL
Unit Size:
Each
Description
- Extracts crystals, granulated and solid substances, thick liquids, and delicate materials
- Features a spoon end and a spatula end
- Made of polished 304 grade stainless steel
- Flameproof, autoclavable, and impervious to boiling water
Specifications
Specifications
| Quantity | 1 |
| Autoclavable | Autoclavable |
| Capacity (Metric) | 10 mL |
| Color | Stainless Steel |
| Product Type | Spoon and Spatula Sampler |
| Finish | None |
| Material | Stainless Steel |
| Length (Metric) | 18 cm |