Learn More
Invitrogen™ eBioscience™ ELISA Wash Buffer, 1L packets
Click to view available options
Quantity:
10 Packets
Unit Size:
Pack of 10
Description
The eBioscience ELISA Wash Buffer Powder is a dried powder formulation of phosphate buffered saline with 0.05% Tween-20. Each packet is reconstituted in 1 Liter of distilled, deionized water, resulting in 1x PBS/Tween-20 buffer (pH 7.4).
Specifications
Specifications
| Product Type | Wash Buffer |
| Quantity | 10 Packets |
| Form | Powder |
| For Use With (Application) | ELISA |
| Product Line | eBioscience |