missing translation for 'onlineSavingsMsg'
Learn More

Elbasvir, MedChemExpress

Product Code. 16479377
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16479377

Brand: Medchem Express HY15789

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Elbasvir (MK-8742) is a hepatitis C virus nonstructural protein 5A (HCV NS5A) inhibitor with EC50s of 4, 3 and 3 nM against genotype 1a, 1b, and 2a, respectively.

Elbasvir (MK-8742) is a hepatitis C virus nonstructural protein 5A (HCV NS5A) inhibitor with EC50s of 4, 3 and 3 nM against genotype 1a, 1b, and 2a, respectively.

Specifications

Chemical Name or Material Elbasvir
CAS 1370468-36-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C49H55N9O7
Quantity 5 mg
Synonym MK-8742
Solubility Information DMSO : ≥ 50 mg/mL (56.69 mM)
SMILES O=C([C@H](C(C)C)NC(OC)=O)N(CCC1)[C@@H]1C(N2)=NC=C2C(C=C3)=CC4=C3C5=CC6=CC(C7=CN=C([C@H]8N(C([C@H](C(C)C)NC(OC)=O)=O)CCC8)N7)=CC=C6N5[C@H](C9=CC=CC=C9)O4
Molecular Weight (g/mol) 882.02
Formula Weight 882.02
Percent Purity 95.67%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.