Learn More
Eight elements solution - Ge, Hf, Mo Sb Sn, Te, W, Zr for ICP/MS 10mg/L, Fisher chemical™
Product Code. 12946203 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
Unit Size:
100mL
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12946203 250 mL 250mL
12926203 50 mL 50mL
12936203 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12946203 Supplier Fisher Chemical Supplier No. MS 011 250
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Chemical Name or Material Eight Elements
Packaging Bottle
Analyte or Component Names Ge,Hf,Mo,Sb,Sn,Te,W,Zr
Physical Form Liquid
Concentration 10 mg/L
Type ICP/MS Calibration Multi-element Standard Solution
Element Ge, Hf, Mo, Sb, Sn, Te, W, Zr
Solvent or Matrix Nitric Acid/Trace Hydrogen Fluoride
Quantity 250 mL
compliance-icons
Product Identifier
  • Multi-element Standard Solution for ICP. 8 Components in HNO3 2%; HF 0.1% (MS 011)
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • EUH210-Safety data sheet available on request.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501-Dispose of contents/container to .